Zinc(II) acetylacetonate monohydrate 98%

Supplier: Ambeed
Warning

A1150334-100G A1150334-500G A1150334-25G
76674-250EA 34 USD
76674-250 76674-252 77620-112
Zinc(II) acetylacetonate monohydrate 98%
Zinc(II) acetylacetonate monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₄Zn·1H₂O
MDL Number: MFCD00149060
CAS Number: 14363-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR