Indium(III) sulfide 99.99% (trace metals basis)
Supplier: Ambeed
|
Warning
|
Synonyms:
Diindium trisulphide, Indium trisulphide
A1150330-1G
A1150330-25G
A1150330-5G
76674-244EA
20.16
USD
76674-244
76674-246
76674-248
Indium(III) sulfide 99.99% (trace metals basis)
Indium(III) sulphide
Precaución: for Research Use Only
Formula:
In₂S₃ MW: 325.84 g/mol |
MDL Number:
MFCD00011061 CAS Number: 12030-24-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...