1-Hexylpyridinium trifluoromethanesulphonate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hexylpyridin-1-ium trifluoromethanesulphonate

A1150288-1G A1150288-25G A1150288-5G
76674-226EA 11 USD
76674-226 76674-228 77865-959
1-Hexylpyridinium trifluoromethanesulphonate 98%
1-Hexylpyridinium trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₁₂H₁₈F₃NO₃S
MW: 313.3364 g/mol
MDL Number: MFCD03427623
CAS Number: 623167-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR