(R)-tert-Butyl (5,6-diamino-6-oxohexyl)carbamate 97%

Supplier: Ambeed
Warning

A1150280-1G A1150280-250MG
76674-222EA 51 USD
76674-222 76674-224
(R)-tert-Butyl (5,6-diamino-6-oxohexyl)carbamate 97%
(R)-tert-Butyl (5,6-diamino-6-oxohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₃N₃O₃
CAS Number: 96138-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR