N-(4-((2,6-Dimethylphenyl)amino)pent-3-en-2-ylidene)-2,6-dimethylaniline 98%

Supplier: Ambeed
Warning

A1150257-100MG A1150257-250MG A1150257-5G A1150257-1G
76674-206EA 28 USD
76674-206 76674-208 76674-210 77620-106
N-(4-((2,6-Dimethylphenyl)amino)pent-3-en-2-ylidene)-2,6-dimethylaniline 98%
N-(4-((2,6-Dimethylphenyl)amino)pent-3-en-2-ylidene)-2,6-dimethylaniline

Precaución: for Research Use Only

Formula: C₂₁H₂₆N₂
MDL Number: MFCD20922892
CAS Number: 267431-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR