4,16-Dibromo[2.2]paracyclophane 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,16-Dibromotricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
A1149914-5G
A1149914-1G
76674-174EA
198.39
USD
76674-174
77620-098
4,16-Dibromo[2.2]paracyclophane 98%
4,16-Dibromo[2.2]paracyclophane
Precaución: for Research Use Only
Formula:
C₁₆H₁₄Br₂ MW: 366.1 g/mol |
MDL Number:
MFCD09953451 CAS Number: 96392-77-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...