1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 98%

Supplier: Ambeed
Warning

Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium dibutyl phosphate

A1149903-100G A1149903-25G A1149903-5G
76674-168EA 136 USD
76674-168 76674-170 76674-172
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate 98%
1-n-Butyl-3-methylimidazolium di-n-butyl phosphate

Precaución: for Research Use Only

Formula: C₁₆H₃₃N₂O₄P
MW: 348.424 g/mol
MDL Number: MFCD09265043
CAS Number: 663199-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR