Tributylhexylphosphonium bromide 98+%

Supplier: Ambeed
Warning

Synonyms: Tributyl(hexyl)phosphonium bromide, Hexyltributylphosphonium bromide

A1149881-1G A1149881-25G A1149881-5G
76674-156EA 8 USD
76674-156 76674-158 77865-950
Tributylhexylphosphonium bromide 98+%
Tributylhexylphosphonium bromide

Precaución: for Research Use Only

Formula: C₁₈H₄₀BrP
MW: 367.40 g/mol
MDL Number: MFCD00134170
CAS Number: 105890-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR