(S)-N-(1-cyclopropyl-2,2,2-trifluoroethyl)-4-(6-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidin-3-yl)thiophene-2-carboxamide 99%

Supplier: Ambeed
Warning

Synonyms: ULK-101

A1149867-10MG A1149867-1MG A1149867-5MG
76674-150EA 636.54 USD
76674-150 76674-152 76674-154
(S)-N-(1-cyclopropyl-2,2,2-trifluoroethyl)-4-(6-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidin-3-yl)thiophene-2-carboxamide 99%
(S)-N-(1-cyclopropyl-2,2,2-trifluoroethyl)-4-(6-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidin-3-yl)thiophene-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₂H₁₆F₄N₄OS
MW: 460.45 g/mol
MDL Number: MFCD31813791
CAS Number: 2443816-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR