1-Octyl-3-methylimidazolium dicyanamide 99%
Supplier: Ambeed
|
Warning
|
A1149827-1G
A1149827-5G
76674-146EA
15.92
USD
76674-146
76674-148
1-Octyl-3-methylimidazolium dicyanamide 99%
1-Octyl-3-methylimidazolium dicyanamide
Precaución: for Research Use Only
Formula:
C₁₄H₂₃N₅ MW: 261.3659 g/mol |
MDL Number:
MFCD19443503 CAS Number: 905972-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...