1-Methyl-3-tetradecyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide 98%

Supplier: Ambeed
Warning

A1149797-25G A1149797-5G
76674-142EA 196.27 USD
76674-142 76674-144
1-Methyl-3-tetradecyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide 98%
1-Methyl-3-tetradecyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide

Precaución: for Research Use Only

Formula: C₂₀H₃₅F₆N₃O₄S₂
MW: 559.6300 g/mol
MDL Number: MFCD17018968
CAS Number: 404001-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR