1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 98%

Supplier: Ambeed
Warning

Synonyms: 3-Dodecyl-1-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide

A1149775-100G A1149775-25G A1149775-5G
76674-136EA 260 USD
76674-136 76674-138 76674-140
1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide 98%
1-Dodecyl-3-methylimidazolium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₈H₃₁F₆N₃O₄S₂
MDL Number: MFCD12022372
CAS Number: 404001-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR