1-Benzyl-3-methylimidazolium dicyanamide 99%

Supplier: Ambeed
Warning

A1149765-1G A1149765-5G A1149765-25G
76674-132EA 9 USD
76674-132 76674-134 77865-947
1-Benzyl-3-methylimidazolium dicyanamide 99%
1-Benzyl-3-methylimidazolium dicyanamide

Precaución: for Research Use Only

Formula: C₁₃H₁₃N₅
MDL Number: MFCD31543876
CAS Number: 958445-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR