N-Octylpyridinium bis(trifluoromethanesulfonyl)imide 99%

Supplier: Ambeed
Warning

A1149750-25G A1149750-5G
76674-128EA 119 USD
76674-128 76674-130
N-Octylpyridinium bis(trifluoromethanesulfonyl)imide 99%
N-Octylpyridinium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₅H₂₂F₆N₂O₄S₂
MW: 472.4666 g/mol
MDL Number: MFCD09038848
CAS Number: 384347-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR