1-Hexyl-3-methyl-1H-imidazol-3-ium hydrogensulfate 98%

Supplier: Ambeed
Danger

A1149732-25G A1149732-1G A1149732-5G
76674-126EA 45.62 USD
76674-126 77620-094 77620-096
1-Hexyl-3-methyl-1H-imidazol-3-ium hydrogensulfate 98%
1-Hexyl-3-methyl-1H-imidazol-3-ium hydrogensulfate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₄S
MW: 264.3418 g/mol
MDL Number: MFCD30183454
CAS Number: 478935-29-4
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR