3-Bromo-11,11-dimethyl-11H-benzo[b]fluorene 98%

Supplier: Ambeed
Warning

A1149658-1G A1149658-5G
76674-116EA 14 USD
76674-116 76674-118
3-Bromo-11,11-dimethyl-11H-benzo[b]fluorene 98%
3-Bromo-11,11-dimethyl-11H-benzo[b]fluorene

Precaución: for Research Use Only

Formula: C₁₉H₁₅Br
MDL Number: MFCD32065724
CAS Number: 1674334-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR