2-Decyl-1-tetradecanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Decyltetradecan-1-ol

A1149601-100G A1149601-25G A1149601-5G
76674-104EA 100 USD
76674-104 76674-106 76674-108
2-Decyl-1-tetradecanol 98%
2-Decyl-1-tetradecanol

Precaución: for Research Use Only

Formula: C₂₄H₅₀O
MW: 354.66 g/mol
MDL Number: MFCD00674096
CAS Number: 58670-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR