4,7-Bis(5-bromo-4-n-octyl-2-thienyl)-2,1,3-benzothiadiazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,7-Bis(5-bromo-4-octylthiophen-2-yl)benzo[c][1,2,5]thiadiazole
A1149571-100MG
A1149571-250MG
76674-092EA
129.43
USD
76674-092
76674-094
4,7-Bis(5-bromo-4-n-octyl-2-thienyl)-2,1,3-benzothiadiazole 98%
4,7-Bis(5-bromo-4-n-octyl-2-thienyl)-2,1,3-benzothiadiazole
Precaución: for Research Use Only
Formula:
C₃₀H₃₈Br₂N₂S₃ MW: 682.64 g/mol |
CAS Number:
457931-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...