4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzoic acid 98%

Supplier: Ambeed
Warning

A1149569-100MG A1149569-1G A1149569-250MG A1149569-5G
76674-084EA 32.89 USD
76674-084 76674-086 76674-088 76674-090
4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzoic acid 98%
4,4'-(1,2-Diphenylethene-1,2-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₀O₄
MW: 420.4560 g/mol
MDL Number: MFCD28901374
CAS Number: 1002339-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR