1,4-Diethynyl-2,5-dimethoxybenzene 98%

Supplier: Ambeed
Warning

A1149561-100MG A1149561-250MG A1149561-5G A1149561-1G
76674-076EA 30 USD
76674-076 76674-078 76674-080 77570-870
1,4-Diethynyl-2,5-dimethoxybenzene 98%
1,4-Diethynyl-2,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
MDL Number: MFCD00846857
CAS Number: 74029-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR