4,4,5,5-Tetramethyl-2-(4-(1,2,2-triphenylvinyl)phenyl)-1,3,2-dioxaborolane 98% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1149541-1G A1149541-250MG A1149541-5G
76674-066EA 93.36 USD
76674-066 76674-068 76674-070
4,4,5,5-Tetramethyl-2-(4-(1,2,2-triphenylvinyl)phenyl)-1,3,2-dioxaborolane 98% stabilized
4,4,5,5-Tetramethyl-2-(4-(1,2,2-triphenylvinyl)phenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₃₂H₃₁BO₂
MW: 458.3984 g/mol
MDL Number: MFCD31697651
CAS Number: 1260865-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR