4,7-Dibromo-2-(2-butyloctyl)-5,6-difluoro-2H-benzo[d][1,2,3]triazole 97%

Supplier: Ambeed
Warning

A1149527-100MG A1149527-250MG
76674-056EA 59.42 USD
76674-056 76674-058
4,7-Dibromo-2-(2-butyloctyl)-5,6-difluoro-2H-benzo[d][1,2,3]triazole 97%
4,7-Dibromo-2-(2-butyloctyl)-5,6-difluoro-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₁₈H₂₅Br₂F₂N₃
MW: 481.2160 g/mol
MDL Number: MFCD31697652
CAS Number: 1283099-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR