4-(1,2,2-Triphenylvinyl)aniline 98%

Supplier: Ambeed
Warning

A1149517-100MG A1149517-1G A1149517-250MG A1149517-5G A1149517-25G
76674-048EA 15.92 USD
76674-048 76674-050 76674-052 76674-054 77620-086
4-(1,2,2-Triphenylvinyl)aniline 98%
4-(1,2,2-Triphenylvinyl)aniline

Precaución: for Research Use Only

Formula: C₂₆H₂₁N
MW: 347.4517 g/mol
MDL Number: MFCD31697689
CAS Number: 919789-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR