(E)-4,4,4-Trifluorobut-2-enoic acid 97%
Supplier: Ambeed
|
Danger
|
A1149514-1G
A1149514-25G
A1149514-5G
76674-042EA
15.92
USD
76674-042
76674-044
76674-046
(E)-4,4,4-Trifluorobut-2-enoic acid 97%
(E)-4,4,4-Trifluorobut-2-enoic acid
Precaución: for Research Use Only
Formula:
C₄H₃F₃O₂ MW: 140.0606 g/mol |
MDL Number:
MFCD02181180 CAS Number: 406-94-0 UN: 3261 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...