1,1,2,2-Tetrakis(4-ethynylphenyl)ethene 98%

Supplier: Ambeed
Warning

A1149510-100MG A1149510-250MG A1149510-1G
76674-038EA 80.63 USD
76674-038 76674-040 77566-046
1,1,2,2-Tetrakis(4-ethynylphenyl)ethene 98%
1,1,2,2-Tetrakis(4-ethynylphenyl)ethene

Precaución: For research use only.

Formula: C₃₄H₂₀
MW: 428.5226 g/mol
MDL Number: MFCD30470530
CAS Number: 4863-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR