1-Methyl-3-tetradecyl-1H-imidazol-3-ium hexafluorophosphate(V) 98%
Supplier: Ambeed
|
Warning
|
A1149504-100G
A1149504-1G
A1149504-25G
A1149504-5G
76674-030EA
439.22
USD
76674-030
76674-032
76674-034
76674-036
1-Methyl-3-tetradecyl-1H-imidazol-3-ium hexafluorophosphate(V) 98%
1-Methyl-3-tetradecyl-1H-imidazol-3-ium hexafluorophosphate(V)
Precaución: for Research Use Only
Formula:
C₁₈H₃₅F₆N₂P MW: 424.4481 g/mol |
CAS Number:
219947-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...