Ethyl 4-((4-hydroxyphenyl)diazenyl)benzoate 98%

Supplier: Ambeed
Warning

A1149487-1G A1149487-250MG A1149487-5G
76674-004EA 13 USD
76674-004 76674-006 76674-008
Ethyl 4-((4-hydroxyphenyl)diazenyl)benzoate 98%
Ethyl 4-((4-hydroxyphenyl)diazenyl)benzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₄N₂O₃
MDL Number: MFCD30730201
CAS Number: 4418-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR