4-(1,2,2-Triphenylvinyl)benzoic acid 98%

Supplier: Ambeed
Warning

A1149480-1G A1149480-250MG A1149480-5G
76673-998EA 24 USD
76673-998 76674-000 76674-002
4-(1,2,2-Triphenylvinyl)benzoic acid 98%
4-(1,2,2-Triphenylvinyl)benzoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₀O₂
MW: 376.4465 g/mol
MDL Number: MFCD31697666
CAS Number: 197153-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR