2-(4-Amino-1-oxoisoindolin-2-yl)pentanedioic acid 95%

Supplier: Ambeed
Warning

A1149419-100MG A1149419-250MG
76673-988EA 303.42 USD
76673-988 76673-990
2-(4-Amino-1-oxoisoindolin-2-yl)pentanedioic acid 95%
2-(4-Amino-1-oxoisoindolin-2-yl)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₅
MW: 278.2607 g/mol
MDL Number: MFCD32644495
CAS Number: 295357-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR