(4-((4-Chloro-3-(trifluoromethyl)phenyl)sulfonyl)piperazin-1-yl)(4-(5-methyl-1H-pyrazol-1-yl)phenyl)methanone ≥99%

Supplier: Ambeed
Warning

Synonyms: Smurf1-IN-A01

A1149393-10MG A1149393-1MG A1149393-25MG A1149393-50MG A1149393-5MG
76673-978EA 110.01 USD
76673-978 76673-980 76673-982 76673-984 76673-986
(4-((4-Chloro-3-(trifluoromethyl)phenyl)sulfonyl)piperazin-1-yl)(4-(5-methyl-1H-pyrazol-1-yl)phenyl)methanone ≥99%
(4-((4-Chloro-3-(trifluoromethyl)phenyl)sulfonyl)piperazin-1-yl)(4-(5-methyl-1H-pyrazol-1-yl)phenyl)methanone

Precaución: for Research Use Only

Formula: C₂₂H₂₀ClF₃N₄O₃S
MDL Number: MFCD09320899
CAS Number: 1007647-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR