(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-dimethoxy-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole 97%

Supplier: Ambeed
Warning

A1149371-100MG A1149371-250MG A1149371-1G
76673-974EA 119.89 USD
76673-974 77595-862 77620-080
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-dimethoxy-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole 97%
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-dimethoxy-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole

Precaución: for Research Use Only

Formula: C₂₄H₃₂O₄P₂
MW: 446.46 g/mol
MDL Number: MFCD31707600
CAS Number: 1202033-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR