1-Ethyl-3-vinyl-1H-imidazol-3-ium tetrafluoroborate 98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1149347-25G A1149347-5G
76673-972EA 49 USD
76673-972 77620-078
1-Ethyl-3-vinyl-1H-imidazol-3-ium tetrafluoroborate 98% stabilized
1-Ethyl-3-vinyl-1H-imidazol-3-ium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₇H₁₁BF₄N₂
MDL Number: MFCD28142374
CAS Number: 936030-51-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR