Benzamidine hydrochloride dihydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzene-1-carboximidamide hydrochloride dihydrate, Phenylamidine hydrochloride dihydrate
A1149301-25G
A1149301-500G
76673-958EA
5.31
USD
76673-958
76673-960
Benzamidine hydrochloride dihydrate 98%
Benzamidine hydrochloride dihydrate
Precaución: for Research Use Only
Formula:
C₇H₉ClN₂·2H₂O MW: 192.64 g/mol |
MDL Number:
MFCD00066285 CAS Number: 74222-83-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...