(E)-3-(4-(Dimethylamino)phenyl)acrylaldehyde 98%

Supplier: Ambeed
Warning

A1149299-25G A1149299-5G
76673-956EA 99.73 USD
76673-956 77566-044
(E)-3-(4-(Dimethylamino)phenyl)acrylaldehyde 98%
(E)-3-(4-(Dimethylamino)phenyl)acrylaldehyde

Precaución: For research use only.

Formula: C₁₁H₁₃NO
MW: 175.2270 g/mol
MDL Number: MFCD00007002
CAS Number: 20432-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR