(E)-4,4'-(Ethene-1,2-diyl)dibenzoic acid 98%

Supplier: Ambeed
Warning

A1149289-1G A1149289-250MG A1149289-5G
76673-950EA 36.08 USD
76673-950 76673-952 76673-954
(E)-4,4'-(Ethene-1,2-diyl)dibenzoic acid 98%
(E)-4,4'-(Ethene-1,2-diyl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₄
MW: 268.2641 g/mol
MDL Number: MFCD00013994
CAS Number: 74299-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR