rel-Di-tert-butyl((2R,3R)-3-(tert-butyl)-4-(dimethylamino)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)phosphine oxide 97%

Supplier: Ambeed
Warning

A1149281-250MG A1149281-100MG A1149281-1G
76673-944EA 273.72 USD
76673-944 77570-860 77620-074
rel-Di-tert-butyl((2R,3R)-3-(tert-butyl)-4-(dimethylamino)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)phosphine oxide 97%
rel-Di-tert-butyl((2R,3R)-3-(tert-butyl)-4-(dimethylamino)-2,3-dihydrobenzo[d][1,3]oxaphosphol-2-yl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₁H₃₇NO₂P₂
MW: 397.4715 g/mol
MDL Number: MFCD31707612
CAS Number: 1788085-47-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR