5-Chloro-2H-benzo[d][1,2,3]triazole 98%

Supplier: Ambeed
Warning

A1149129-100G A1149129-25G A1149129-5G
76673-928EA 43.5 USD
76673-928 76673-930 76673-932
5-Chloro-2H-benzo[d][1,2,3]triazole 98%
5-Chloro-2H-benzo[d][1,2,3]triazole

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5691 g/mol
MDL Number: MFCD00005700
CAS Number: 1185742-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR