4-(((5-Bromo-2-((3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl)amino)pyrimidin-4-yl)amino)methyl)benzenesulfonamide 99+%

Supplier: Ambeed
Warning

Synonyms: GSK8612

A1149118-10MG A1149118-1MG A1149118-25MG A1149118-5MG
76673-920EA 71.09 USD
76673-920 76673-922 76673-924 76673-926
4-(((5-Bromo-2-((3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl)amino)pyrimidin-4-yl)amino)methyl)benzenesulfonamide 99+%
4-(((5-Bromo-2-((3-methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl)amino)pyrimidin-4-yl)amino)methyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₇H₁₇BrF₃N₇O₂S
MW: 520.33 g/mol
MDL Number: MFCD32062739
CAS Number: 2361659-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR