4-((3,4-Dimethylphenyl)amino)-7-(pyridin-4-yl)quinoline-3-carboxamide 99%

Supplier: Ambeed
Warning

Synonyms: cFMS Receptor Inhibitor II

A1149117-10MG A1149117-1MG A1149117-5MG
76673-914EA 64.72 USD
76673-914 76673-916 76673-918
4-((3,4-Dimethylphenyl)amino)-7-(pyridin-4-yl)quinoline-3-carboxamide 99%
4-((3,4-Dimethylphenyl)amino)-7-(pyridin-4-yl)quinoline-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₀N₄O
MW: 368.43 g/mol
MDL Number: MFCD28138115
CAS Number: 959860-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR