3-(Benzylamino)-4-(cyclohexylamino)-N-(2-(piperazin-1-yl)ethyl)benzenesulfonamide hydrochloride 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
UAMC-3203 HCl
A1149114-10MG
A1149114-1MG
A1149114-50MG
A1149114-5MG
76673-888EA
59.42
USD
76673-888
76673-890
76673-892
76673-894
3-(Benzylamino)-4-(cyclohexylamino)-N-(2-(piperazin-1-yl)ethyl)benzenesulfonamide hydrochloride 99%
3-(Benzylamino)-4-(cyclohexylamino)-N-(2-(piperazin-1-yl)ethyl)benzenesulfonamide hydrochloride
Precaución: for Research Use Only
Formula:
C₂₅H₃₈ClN₅O₂S MW: 508.12 g/mol |
MDL Number:
MFCD32062758 CAS Number: 2271358-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...