(4-(4-(6-Fluorobenzo[d]oxazol-2-yl)benzoyl)piperazin-1-yl)(1-hydroxycyclopropyl)methanone 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
FT113
A1149113-10MG
A1149113-1MG
A1149113-25MG
A1149113-50MG
A1149113-5MG
76673-878EA
70.02
USD
76673-878
76673-880
76673-882
76673-884
76673-886
(4-(4-(6-Fluorobenzo[d]oxazol-2-yl)benzoyl)piperazin-1-yl)(1-hydroxycyclopropyl)methanone 99%
(4-(4-(6-Fluorobenzo[d]oxazol-2-yl)benzoyl)piperazin-1-yl)(1-hydroxycyclopropyl)methanone
Precaución: for Research Use Only
Formula:
C₂₂H₂₀FN₃O₄ MW: 409.41 g/mol |
MDL Number:
MFCD31813811 CAS Number: 1630808-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...