JNJ-61432059 99%

Supplier: Ambeed
Warning

A1149111-10MG A1149111-1MG A1149111-5MG
76673-862EA 168.69 USD
76673-862 76673-864 76673-866
JNJ-61432059 99%
JNJ-61432059

Precaución: for Research Use Only

Formula: C₂₅H₂₂FN₅O₂
MW: 443.47 g/mol
CAS Number: 2035814-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR