rel-(2R,3R)-2-[Bis(1,1-dimethylethyl)phosphinyl]-3-(1,1-dimethylethyl)-2,3-dihydro-4-methoxy-1,3-benzoxaphosphole 97%

Supplier: Ambeed
Warning

A1149087-100MG A1149087-250MG
76673-850EA 74.27 USD
76673-850 76673-852
rel-(2R,3R)-2-[Bis(1,1-dimethylethyl)phosphinyl]-3-(1,1-dimethylethyl)-2,3-dihydro-4-methoxy-1,3-benzoxaphosphole 97%
rel-(2R,3R)-2-[Bis(1,1-dimethylethyl)phosphinyl]-3-(1,1-dimethylethyl)-2,3-dihydro-4-methoxy-1,3-benzoxaphosphole

Precaución: for Research Use Only

Formula: C₂₀H₃₄O₃P₂
MW: 384.4297 g/mol
MDL Number: MFCD31707611
CAS Number: 1788085-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR