2-((3,5-Dibromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol 98%

Supplier: Ambeed
Warning

A1148997-1G A1148997-250MG A1148997-5G
76673-822EA 68.96 USD
76673-822 76673-824 76673-826
2-((3,5-Dibromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol 98%
2-((3,5-Dibromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol

Precaución: for Research Use Only

Formula: C₁₅H₁₆Br₂N₄O
MW: 428.1217 g/mol
MDL Number: MFCD17019234
CAS Number: 14337-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR