3-(3-(Dimethoxymethyl)-1H-pyrazol-5-yl)pyridine 97%

Supplier: Ambeed
Warning

A1148945-100MG A1148945-1G A1148945-250MG A1148945-5G
76673-808EA 40 USD
76673-808 76673-810 76673-812 76673-814
3-(3-(Dimethoxymethyl)-1H-pyrazol-5-yl)pyridine 97%
3-(3-(Dimethoxymethyl)-1H-pyrazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃O₂
CAS Number: 1680191-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR