(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-diphenyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole 97%
Supplier: Ambeed
|
Warning
|
A1148895-100MG
A1148895-250MG
A1148895-25MG
A1148895-50MG
A1148895-1G
76673-804EA
201
USD
76673-804
77620-058
77620-060
77620-062
77783-310
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-diphenyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole 97%
(2S,2'S,3S,3'S)-3,3'-Di-tert-butyl-4,4'-diphenyl-2,2',3,3'-tetrahydro-2,2'-bibenzo[d][1,3]oxaphosphole
Precaución: for Research Use Only
Formula:
C₃₄H₃₆O₂P₂ |
MDL Number:
MFCD31707602 CAS Number: 1202033-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...