5-Bromo-4-methyl-thiazol-2-ylamine hydrobromide 97%

Supplier: Ambeed
Warning

A1148251-5G A1148251-25G
76673-770EA 56.23 USD
76673-770 77620-042
5-Bromo-4-methyl-thiazol-2-ylamine hydrobromide 97%
5-Bromo-4-methyl-thiazol-2-ylamine hydrobromide

Precaución: for Research Use Only

Formula: C₄H₆Br₂N₂S
MW: 273.9768 g/mol
MDL Number: MFCD19441981
CAS Number: 79247-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR