3,4-Dichloroisocoumarin 98%

Supplier: Ambeed
Danger

A1148102-10MG A1148102-1MG A1148102-5MG
76673-762EA 82 USD
76673-762 76673-764 76673-766
3,4-Dichloroisocoumarin 98%
3,4-Dichloroisocoumarin

Precaución: for Research Use Only

Formula: C₉H₄Cl₂O₂
MDL Number: MFCD00036960
CAS Number: 51050-59-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR