1-(7-Methoxy-4-methylquinazolin-2-yl)guanidine 98%

Supplier: Ambeed
Warning

A1148094-10MG A1148094-1MG A1148094-5MG
76673-756EA 43 USD
76673-756 76673-758 76673-760
1-(7-Methoxy-4-methylquinazolin-2-yl)guanidine 98%
1-(7-Methoxy-4-methylquinazolin-2-yl)guanidine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₅O
MDL Number: MFCD00475732
CAS Number: 37836-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR