2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%

Supplier: Ambeed
Warning

A1148048-1G A1148048-250MG A1148048-25G A1148048-5G
76673-740EA 56 USD
76673-740 76673-742 76673-744 76673-746
2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione 98%
2-(4-Iodophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Precaución: for Research Use Only

Formula: C₁₁H₁₁BINO₄
MDL Number: MFCD11215234
CAS Number: 1287221-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR